6-Chloro-2-fluoro-3-methylbenzoylchloride 97%

Danger

Synonyms: 6-Chloro-2-fluoro-3-toluoyl chloride6-Chloro-2-fluoro-3-methylbenzene-1-carbonyl chloride

B23374-03 B23374-06 B23374-03 B23374-06
AAB23374-03EA 239.28 USD
CAAAB23374-03 CAAAB23374-06 AAB23374-03 AAB23374-06
6-Chloro-2-fluoro-3-methylbenzoylchloride 97%
6-Chloro-2-fluoro-3-toluoyl chloride
Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
Melting Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD01631371
CAS Number: 261762-81-6
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR