2-Chloro-6-fluoro-m-toluic acid 97%
|
Warning
|
Synonyms:
2-Chloro-6-fluoro-3-methylbenzoic acid
B23281-03
B23281-06
B23281-06
B23281-03
AAB23281-06EA
162.06
USD
CAAAB23281-03
CAAAB23281-06
AAB23281-06
AAB23281-03
2-Chloro-6-fluoro-m-toluic acid 97%
2-Chloro-6-fluoro-m-toluic acid
Formula:
C₈H₆ClFO₂ MW: 188.59 g/mol Melting Pt: 116…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631359 CAS Number: 32890-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...