Hexachloroacetone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexachloro-2-propanone

B23259-30 B23259-0B B23259-0B B23259-30
AAB23259-0BEA 111.99 USD
AAB23259-0B CAAAB23259-30 AAB23259-30 CAAAB23259-0B
Hexachloroacetone 99%
Hexachloroacetone
Formula: C₃Cl₆O
MW: 264.75 g/mol
Boiling Pt: 203…205 °C
Melting Pt: –2 °C
Density: 1.748
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000796
CAS Number: 116-16-5
EINECS: 204-129-5
UN: 2661
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR