4-Methyl-3-(trifluoromethyl)benzonitrile 97+%
|
Danger
|
B23245-06
B23245-03
B23245-06
B23245-03
AAB23245-03EA
58.24
USD
CAAAB23245-03
CAAAB23245-06
AAB23245-03
AAB23245-06
4-Methyl-3-(trifluoromethyl)benzonitrile 97+%
4-Methyl-3-(trifluoromethyl)benzonitrile
Formula:
C₉H₆F₃N Melting Pt: 60…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631597 CAS Number: 261952-06-1 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...