Methyl-3-methoxy-4-nitrobenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Methoxy-4-nitrobenzoic acid methyl esterMethyl 3-methoxy-4-nitrobenzoateMethyl 3-methoxy-4-nitrobenzenecarboxylate

B23196-03 B23196-03 B23196-06 B23196-06
AAB23196-06EA 210 USD
CAAAB23196-03 AAB23196-06 CAAAB23196-06 AAB23196-03
Methyl-3-methoxy-4-nitrobenzoate 98%
Methyl 3-methoxy-4-nitrobenzoate
Formula: C₉H₉NO₅
MW: 211.17 g/mol
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00051967
CAS Number: 5081-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR