5-Chloro-2-fluorobenzonitrile 97+%

Danger

B23120-03 B23120-06 B23120-03 B23120-06
AAB23120-03EA 55.95 USD
CAAAB23120-06 CAAAB23120-03 AAB23120-03 AAB23120-06
5-Chloro-2-fluorobenzonitrile 97+%
5-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01631434
CAS Number: 57381-34-7
EINECS: 260-710-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR