3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride 97%
|
Danger
|
B23069-06
B23069-06
B23069-03
B23069-03
AAB23069-03EA
67.61
USD
AAB23069-03
CAAAB23069-06
AAB23069-06
CAAAB23069-03
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride 97%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride
Formula:
C₈H₂Cl₂F₄O MW: 261 g/mol Melting Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631511 CAS Number: 261763-03-5 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...