Hexamethylcyclotrisiloxane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-trioxatrisilinane

B23028-14 B23028-36 B23028-14 B23028-22 B23028-22 B23028-36
AAB23028-14EA 39.79 USD
CAAAB23028-22 CAAAB23028-36 CAAAB23028-14 AAB23028-14 AAB23028-36 AAB23028-22
Hexamethylcyclotrisiloxane 97%
Hexamethylcyclotrisiloxane
Formula: C₆H₁₈O₃Si₃
MW: 222.46 g/mol
Boiling Pt: 134 °C
Melting Pt: 64…66 °C
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005943
CAS Number: 541-05-9
EINECS: 208-765-4
UN: 1325
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR