2-Bromobenzaldehyde oxime 96%

Warning

Synonyms: 2-Bromobenzaldoxime

B23023-06 B23023-14 B23023-03 B23023-06 B23023-03 B23023-14
AAB23023-06EA 96.9 USD
AAB23023-06 AAB23023-03 AAB23023-14 CAAAB23023-06 CAAAB23023-14 CAAAB23023-03
2-Bromobenzaldehyde oxime 96%
2-Bromobenzaldehyde oxime
Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00082734
CAS Number: 34158-72-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR