3-Fluoro-4-(trifluoromethyl)benzylamine 97%

Danger

B23012-06 B23012-03 B23012-06 B23012-03
AAAB23012-03EA 83.4 USD
AAAB23012-03 CAAAAB23012-06 AAAB23012-06 CAAAAB23012-03
3-Fluoro-4-(trifluoromethyl)benzylamine 97%
3-Fluoro-4-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
MW: 193.14 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00236287
CAS Number: 235106-09-9
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR