sec-Butylbenzene 99%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2-Phenylbutane

B22962-AC B22962-AC
AAB22962-ACEA 41.65 USD
CAAAB22962-AC AAB22962-AC
sec-Butylbenzene 99%
sec-Butylbenzene
Formula: C₆H₅CH(CH₃)CH₂CH₃
MW: 134.22 g/mol
Boiling Pt: 173…174 °C
Melting Pt: –75 °C
Density: 0.862
Flash Pt: 52 °C(125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009329
CAS Number: 135-98-8
EINECS: 205-227-0
UN: 2709
ADR: "3",III
Merck Index: 13,01549
https://us.vwr.com/stibo/web/std.lang.all/58/12/765812.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR