α-Bromo-3-chloro-2,6-difluorotoluene 97%

Danger

Synonyms: 3-Chloro-2,6-difluorobenzyl bromide3-Chloro-2,6-difluorobenzylbromide

B22920-03 B22920-06 B22920-06 B22920-03
AAB22920-03EA 99.56 USD
AAB22920-03 CAAAB22920-03 AAB22920-06 CAAAB22920-06
α-Bromo-3-chloro-2,6-difluorotoluene 97%
α-Bromo-3-chloro-2,6-difluorotoluene
Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
Melting Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD01631326
CAS Number: 261762-47-4
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR