2,3-Difluoroanisole 97+%
|
Warning
|
B22916-06
B22916-03
B22916-03
B22916-06
AAB22916-06EA
189.81
USD
CAAAB22916-03
AAB22916-06
CAAAB22916-06
AAB22916-03
2,3-Difluoroanisole 97+%
2,3-Difluoroanisole
Formula:
C₇H₆F₂O MW: 144.12 g/mol Flash Pt: 62 °C(144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01631562 CAS Number: 134364-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...