Cycloheptane 99%

Supplier: Thermo Scientific Chemicals

Danger

B22902-06 B22902-14 B22902-06 B22902-14
AAB22902-06EA 37.3 USD
AAB22902-06 AAB22902-14 CAAAB22902-14 CAAAB22902-06
Cycloheptane 99%
Cycloheptane
Formula: C₇H₁₄
Boiling Pt: 118…119 °C
Melting Pt: –12 °C
Density: 0.811
Flash Pt: 6 °C(43 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004147
CAS Number: 291-64-5
EINECS: 206-030-2
UN: 2241
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR