1,4,7,10-Tetraazacyclododecane tetrahydrochloride 98%

Warning

Synonyms: Cyclen tetrahydrochloride

B22268-06 B22268-01 B22268-06 B22268-01 B22268-03 B22268-03
AAAB22268-03EA 177.08 USD
CAAAAB22268-06 AAAB22268-03 AAAB22268-01 CAAAAB22268-03 CAAAAB22268-01 AAAB22268-06
1,4,7,10-Tetraazacyclododecane tetrahydrochloride 98%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride
Formula: C₈H₂₄Cl₄N₄
Melting Pt: >290 °C
Storage Temperature: Ambient
MDL Number: MFCD00068484
CAS Number: 10045-25-7
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR