3,5-Dichlorobenzamide 97%

B21959-22 B21959-06 B21959-14 B21959-22 B21959-06 B21959-14
AAAB21959-06EA 199.13 USD
CAAAAB21959-22 CAAAAB21959-06 AAAB21959-06 AAAB21959-22 CAAAAB21959-14 AAAB21959-14
3,5-Dichlorobenzamide 97%
3,5-Dichlorobenzamide
Formula: C₇H₅Cl₂NO
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00057644
CAS Number: 5980-23-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR