meta-Phosphoric acid 39 - 43% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Metaphosphoric acid

B20932-36 B20932-22 B20932-22 B20932-36
AAAB20932-36EA 389.27 USD
AAAB20932-36 CAAAAB20932-36 AAAB20932-22 CAAAAB20932-22
meta-Phosphoric acid 39 - 43% stabilized
meta-Phosphoric acid
Formula: (HPO₃)n
MW: 79.98 g/mol
Melting Pt: 21 °C
Density: 2.2
Storage Temperature: Ambient
MDL Number: MFCD00011351
CAS Number: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR