3-Methylcinnamic acid predominantly trans 98%

Warning

Synonyms: 3-(m-Tolyl)acrylic acid

B20914-04 B20914-09 B20914-04 B20914-09
AAAB20914-09EA 192.33 USD
AAAB20914-09 CAAAAB20914-09 CAAAAB20914-04 AAAB20914-04
3-Methylcinnamic acid predominantly trans 98%
3-Methylcinnamic acid
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00016844
CAS Number: 3029-79-6
EINECS: 221-199-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR