2,3,4,5,6-Pentachloroaniline 96%

Danger

Synonyms: Pentachloroaniline

B20327-14 B20327-06 B20327-14 B20327-06
AAAB20327-06EA 79.39 USD
CAAAB20327-14 AAAB20327-06 CAAAAB20327-06 AAB20327-14
2,3,4,5,6-Pentachloroaniline 96%
2,3,4,5,6-Pentachloroaniline
Formula: C₆H₂Cl₅N
MW: 265.35 g/mol
Melting Pt: 231…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00014769
CAS Number: 527-20-8
EINECS: 208-410-3
UN: 2018
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR