1-Acetyl-5-nitroindoline 98%, Technical Grade

Synonyms: 1-(5-Nitroindolin-1-yl)ethanone

B20212-88 B20212-88 B20212-09 B20212-09
AAB20212-88EA 56.12 USD
CAAAB20212-09 AAB20212-88 AAB20212-09 CAAAB20212-88
1-Acetyl-5-nitroindoline 98%, Technical Grade
1-Acetyl-5-nitroindoline
Formula: C₁₀H₁₀N₂O₃
MW: 206.2 g/mol
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00005811
CAS Number: 33632-27-8
EINECS: 251-609-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR