Sodium molybdate(VI) dihydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium molybdenum oxide dihydrateMolybdic acid sodium salt dihydrateSodium molybdate dihydrateMolybdic acid, sodium salt dihydratedi-Sodium molybdate dihydrateMolybdic acid disodium salt dihydrate

A19222-22 A19222-0E A19222-36
AAA19222-0EEA 608.99 USD
AAA19222-0E AAA19222-36 AAA19222-22
Sodium molybdate(VI) dihydrate 98%
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Melting Pt: 100 °C (–H₂O of crystall.)
Density: 2.71
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR