Potassium dichromate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18722-36
A18722-0E
AAAA18722-0EEA
221.99
USD
AAAA18722-0E
AAAA18722-36
Potassium dichromate 99%
Potassium dichromate
Formula:
K₂Cr₂O₇ MW: 294.18 g/mol Melting Pt: 398 °C Density: 2.676 Storage Temperature: Ambient |
MDL Number:
MFCD00011367 CAS Number: 7778-50-9 EINECS: 231-906-6 UN: 3087 ADR: "5.1",III REACH: 01-2119454792-32 Merck Index: 12,07790 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...