Lithium aluminum hydride 97%, powder

Danger

Synonyms: Lithium tetrahydridoaluminate Aluminium lithium hydride

A18116-14 A18116-22 A18116-09
AAA18116-09EA 44.78 USD
CAAAAA18116-14 CAAAAA18116-22 AAA18116-09
Lithium aluminum hydride 97%, powder
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: >125 °C (dec.)
Density: 0.917
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: "4.3",I
Merck Index: 13,00348

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR