Lithium niobate 99%

Synonyms: Lithium niobium oxide

A17805-18 A17805-30 A17805-06 A17805-06 A17805-30 A17805-18
AAAA17805-18EA 125.21 USD
AAAA17805-18 AAAA17805-06 CAAAAA17805-18 CAAAAA17805-06 AAAA17805-30 CAAAAA17805-30
Lithium niobate 99%
Lithium niobate
Formula: LiNbO₃
MW: 147.85 g/mol
Melting Pt: ca 1275 °C
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00011080
CAS Number: 12031-63-9
EINECS: 234-755-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR