Lithium dihydrogen orthophosphate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium phosphate monobasicLithium dihydrogenorthophosphateLithium dihydrogenphosphateLithium dihydrogen phosphate

A16987-30 A16987-30
AAAA16987-30EA 92.69 USD
AAAA16987-30 CAAAAA16987-30
Lithium dihydrogen orthophosphate 97%
Lithium dihydrogen orthophosphate
Formula: H₂LiO₄P
MW: 103.93 g/mol
Melting Pt: >100 °C
Density: 2,461
Storage Temperature: Ambient
MDL Number: MFCD00016188
CAS Number: 13453-80-0
EINECS: 236-633-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR