Lanolin

Supplier: Thermo Scientific Chemicals

Synonyms: Wool fat

A16902-30 A16902-0B A16902-0B A16902-30
AAAA16902-0BEA 104.46 USD
AAAA16902-0B AAAA16902-30 CAAAAA16902-0B CAAAAA16902-30
Lanolin
Lanolin
Melting Pt: 38…44 °C
Density: 0.932…0.945 g/cm³ (15 °C)
Flash Pt: (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00081740
CAS Number: 8006-54-0
EINECS: 232-348-6
Merck Index: 12,05371

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR