Cobalt(II,III) oxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tricobalt tetraoxide
, Cobaltic tetraoxide
A16121-30
A16121-18
A16121-0B
AAA16121-0BEA
638.99
USD
AAA16121-0B
AAA16121-30
AAA16121-18
Cobalt(II,III) oxide
Cobalt (II,III) oxide
Formula:
Co₃O₄ MW: 240.8 g/mol Melting Pt: 900 °C (dec.) Density: 6.11 Storage Temperature: Ambient |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 ADR: 9,III Merck Index: 13,02454 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...