Methyl anthranilate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Aminobenzoic acid methyl esterMethyl 2-aminobenzoateMethyl 2-Anthranilate2-Aminobenzoic acid methyl ester

A15681-0B A15681-36 A15681-0B A15681-30
AAA15681-0BEA 107 USD
CAAAA15681-0B AAA15681-0B AAA15681-36 AAA15681-30
Methyl anthranilate 99%
Methyl anthranilate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 260 °C
Melting Pt: 24 °C
Density: 1,170
Flash Pt: 123 °C(253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007710
CAS Number: 134-20-3
EINECS: 205-132-4
Merck Index: 12,06099

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR