4-Nitrobenzaldehyde oxime 98%

Danger

Synonyms: 4-Nitrobenzaldoxime

A15525-14 A15525-14 A15525-06 A15525-06
AAA15525-06EA 48.5 USD
CAAAA15525-14 AAA15525-06 AAA15525-14 CAAAA15525-06
4-Nitrobenzaldehyde oxime 98%
4-Nitrobenzaldehyde oxime
Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007377
CAS Number: 1129-37-9
EINECS: 214-445-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR