Barium thiosulfate 98%

Warning

Synonyms: Barium hyposulfiteBarium hyposulphite

A14981-22 A14981-14 A14981-36 A14981-14 A14981-22 A14981-36
AAAA14981-14EA 20.41 USD
AAAA14981-14 AAAA14981-36 CAAAAA14981-22 AAAA14981-22 CAAAAA14981-14 CAAAAA14981-36
Barium thiosulfate 98%
Barium thiosulphate
Formula: BaO₃S₂
MW: 249.46 g/mol
Density: 3.45
Storage Temperature: Ambient
MDL Number: MFCD00040661
CAS Number: 35112-53-9
EINECS: 252-375-7
UN: 1564
ADR: "6.1",III
Merck Index: 12,01028

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR