4-Chloro-2-fluorotoluene 99%

Warning

A14753-22 A14753-30 A14753-30 A14753-14 A14753-14 A14753-22
AAAA14753-14EA 58.08 USD
CAAAAA14753-14 AAAA14753-14 CAAAAA14753-22 AAAA14753-30 AAAA14753-22 CAAAAA14753-30
4-Chloro-2-fluorotoluene 99%
4-Chloro-2-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 158…160 °C
Density: 1.184
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000571
CAS Number: 452-75-5
EINECS: 207-210-3
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR