Hydrazine hydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrazine monohydrate
, Hydrazinium hydroxide
A14005-18
A14005-0E
A14005-36
A14005-18
A14005-30
AAAA14005-0EEA
319.57
USD
AAAA14005-0E
CAAAAA14005-18
AAAA14005-18
AAAA14005-36
AAAA14005-30
Hydrazine hydrate 98+%
Hydrazine hydrate
Formula:
NH₂NH₂·H₂O MW: 32.05 g/mol (anhy) Boiling Pt: 120…121 °C Melting Pt: –52 °C Density: 1.027 Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00149931 CAS Number: 10217-52-4 EINECS: 206-114-9 UN: 2030 ADR: "8",III Merck Index: 13,04790 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...