1-Acetyl-4-piperidone 99%

Synonyms: N-Acetyl-4-piperidone

A13512-06 A13512-14 A13512-06 A13512-14
AAA13512-14EA 156.99 USD
CAAAA13512-14 AAA13512-14 AAA13512-06 CAAAA13512-06
1-Acetyl-4-piperidone 99%
1-Acetyl-4-piperidone
Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 218 °C
Density: 1,146
Flash Pt: 112 °C(234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006190
CAS Number: 32161-06-1
EINECS: 250-937-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR