1-Methyl-4-piperidone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methyl-4-piperidone

A13352-18 A13352-22 A13352-22 A13352-0B A13352-30 A13352-18 A13352-0B A13352-30
AAAA13352-0BEA 705.99 USD
AAAA13352-0B AAAA13352-18 CAAAAA13352-18 AAA13352-22 CAAAAA13352-30 CAAAAA13352-0B CAAAA13352-22 AAAA13352-30
1-Methyl-4-piperidone 98%
1-Methyl-4-piperidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 55…58 °C/10 mm
Density: 0.973
Flash Pt: 60 °C(140 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR