Methyl-4-amino-2-methoxybenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-2-methoxybenzoic acid methyl ester
A13239-06
A13239-03
A13239-06
A13239-03
AAA13239-03EA
15.37
USD
AAA13239-03
CAAAA13239-03
AAA13239-06
CAAAA13239-06
Methyl-4-amino-2-methoxybenzoate 98%
Methyl 4-amino-2-methoxybenzoate
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Melting Pt: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017202 CAS Number: 27492-84-8 EINECS: 248-494-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...