1-Hexyne 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13156-30 A13156-18 A13156-09 A13156-09
AAAA13156-30EA 326.36 USD
AAAA13156-30 AAAA13156-09 AAAA13156-18 CAAAAA13156-09
1-Hexyne 98+%
1-Hexyne
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 69…71 °C
Melting Pt: –132 °C
Density: 0.718
Flash Pt: -21 °C(-5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009504
CAS Number: 693-02-7
EINECS: 211-736-9
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR