Acrylonitrile 99+% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 40 ppm

A13058-0F A13058-AP A13058-0F A13058-AP A13058-AE A13058-AE
AAAA13058-0FEA 94.23 USD
AAAA13058-0F CAAAAA13058-0F AAAA13058-AE CAAAAA13058-AE AAAA13058-AP CAAAAA13058-AP
Acrylonitrile 99+% stabilized
Acrylonitrile
Formula: C₃H₃N
MW: 53.06 g/mol
Boiling Pt: 77…78 °C
Melting Pt: –84 °C
Density: 0.806
Flash Pt: 0 °C(32 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001927
CAS Number: 107-13-1
EINECS: 203-466-5
UN: 1093
ADR: "3",I
Merck Index: 13,00133

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR