1-Iodohexadecane 98% stabilized
Supplier: Thermo Scientific Chemicals
Stabilization:
Stabilised with copper
A12977-14
A12977-22
A12977-30
AAA12977-14EA
43.09
USD
AAA12977-14
AAA12977-22
AAA12977-30
1-Iodohexadecane 98% stabilized
1-Iodohexadecane
Formula:
C₁₆H₃₃I MW: 352.34 g/mol Boiling Pt: 206…207 °C/10 mm Melting Pt: 21…23 °C Density: 1.121 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001089 CAS Number: 544-77-4 EINECS: 208-879-4 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...