2-(4-Toluoyl)benzoic acid 98%

Warning

Synonyms: 2-(4-Methylbenzoyl)benzenecarboxylic acid

A12795-0E A12795-0E
AAA12795-0EEA 494.94 USD
CAAAA12795-0E AAA12795-0E
2-(4-Toluoyl)benzoic acid 98%
2-(4-Toluoyl)benzoic acid
Formula: C₁₅H₁₂O₃
MW: 240.26 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00020287
CAS Number: 85-55-2
EINECS: 201-614-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR