2-Naphthalenethiol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thionaphthol

A12125-18 A12125-09 A12125-18 A12125-30 A12125-30 A12125-09
AAA12125-30EA 1280.35 USD
CAAAA12125-18 CAAAA12125-09 AAA12125-30 AAA12125-09 AAA12125-18 CAAAA12125-30
2-Naphthalenethiol 98+%
2-Naphthalenethiol
Formula: C₁₀H₈S
MW: 160.24 g/mol
Boiling Pt: 286 °C
Melting Pt: 79…83 °C
Density: 1.55
Storage Temperature: Ambient
MDL Number: MFCD00004086
CAS Number: 91-60-1
EINECS: 202-082-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR