Sodium thiocyanate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium rhodanide
A11867-30
A11867-0I
A11867-0I
A11867-0B
A11867-30
A11867-0B
AAAA11867-0IEA
365.99
USD
CAAAAA11867-30
AAAA11867-0I
CAAAAA11867-0I
AAAA11867-0B
CAAAAA11867-0B
AAAA11867-30
Sodium thiocyanate 96%
Sodium thiocyanate
Formula:
NaSCN MW: 81.07 g/mol Melting Pt: 287 °C Density: 1.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011123 CAS Number: 540-72-7 EINECS: 208-754-4 Merck Index: 13,09401 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...