3-Iodopropene 97+% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Allyl iodide
, 3-Iodo-1-Propene
, 3-Propenyl iodide
Stabilization:
Stabilised with copper
A11822-30
A11822-18
A11822-18
A11822-30
AAA11822-18EA
121.42
USD
CAAAA11822-18
AAA11822-18
CAAAA11822-30
AAA11822-30
3-Iodopropene 97+% stabilized
3-Iodopropene
Formula:
C₃H₅I MW: 167.98 g/mol Boiling Pt: 101…103 °C Melting Pt: –99 °C Density: 1.832 Flash Pt: 18 °C(64 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001094 CAS Number: 556-56-9 EINECS: 209-130-4 UN: 1723 ADR: "3",II Merck Index: 13,00291 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...