4-Bromo-2-chlorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11527-06 A11527-14 A11527-06 A11527-03 A11527-03 A11527-14
AAAA11527-06EA 82.79 USD
AAAA11527-06 CAAAAA11527-03 CAAAAA11527-14 AAAA11527-14 AAAA11527-03 CAAAAA11527-06
4-Bromo-2-chlorobenzoic acid 98+%
4-Bromo-2-chlorobenzoic acid
Formula: C₇H₄BrClO₂
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00040903
CAS Number: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR