5-Nitro-o-toluonitrile 98%

Warning

Synonyms: 2-Methyl-5-nitrobenzonitrile

A11392-14 A11392-22 A11392-22 A11392-06 A11392-14
AAAA11392-14EA 294.36 USD
CAAAAA11392-22 CAAAAA11392-06 AAAA11392-14 CAAAAA11392-14 AAAA11392-22
5-Nitro-o-toluonitrile 98%
5-Nitro-o-toluonitrile
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00017040
CAS Number: 939-83-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR