2-Chlorobenzhydrazide 98+%
|
Warning
|
Synonyms:
2-Chlorobenzohydrazide
, 2-Chlorobenzoic hydrazide
, 2-Chlorobenzene-1-carbohydrazide
, 2-Chlorobenzoic acid hydrazide
A11361-06
A11361-06
A11361-14
A11361-14
AAAA11361-06EA
30.74
USD
AAAA11361-06
CAAAAA11361-14
AAAA11361-14
CAAAAA11361-06
2-Chlorobenzhydrazide 98+%
2-Chlorobenzhydrazide
Formula:
C₇H₇ClN₂O MW: 170.6 g/mol Melting Pt: 116…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007597 CAS Number: 5814-05-1 EINECS: 227-380-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...