p-Xylene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Dimethylbenzene

A10534-0F A10534-AE A10534-AP
AAAA10534-0FEA 184.1 USD
AAAA10534-0F AAAA10534-AE AAAA10534-AP
p-Xylene 99%
p-Xylene
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 137…138 °C
Melting Pt: 12…13 °C
Density: 0.866
Flash Pt: 27 °C(80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: "3",III
Merck Index: 14,10081
https://us.vwr.com/stibo/web/std.lang.all/87/75/768775.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR