6-Bromohexanenitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromocapronitrile6-Bromohexanonitrile

A10400-09 A10400-04 A10400-09
AAAA10400-09EA 115 USD
CAAAAA10400-09 AAAA10400-09 AAA10400-04
6-Bromohexanenitrile 97%
6-Bromohexanenitrile
Formula: BrH₂C(CH₂)₃CH₂CN
MW: 176.06 g/mol
Boiling Pt: 134…135 °C/15 mm
Density: 1.328
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013834
CAS Number: 6621-59-6
EINECS: 229-570-0
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR