Boron nitride ≥99.5% (metals basis), rods low binder, Length 203mm, Diameter 6.4mm

Supplier: Thermo Scientific Chemicals

45573-KG
AA45573-KGEA 212.99 USD
AA45573-KG
Boron nitride ≥99.5% (metals basis), rods low binder, Length 203mm, Diameter 6.4mm
Boron nitride
Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 13,01336

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR