Gadolinium ≥99.9% (by rare eath metals basis), ingot

Supplier: Thermo Scientific Chemicals

045529.KG
AA45529-KGEA 183.99 USD
AA45529-KG
Gadolinium ≥99.9% (by rare eath metals basis), ingot
Gadolinium
Formula: Gd
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR