Osmic acid 4% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Osmium TetroxideOsmium tetraoxideOsmic Acid AnhydrideOsmium (VIII) oxide

45385-9W 45385-AB
AA45385-ABEA 186.56 USD
AA45385-AB AA45385-9W
Osmic acid 4% in aqueous solution
Osmic acid
Formula: O₄Os
MW: 254.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR